ContactUsLinkComponent
VWR® Measuring Beakers with Handle, Porcelain
VWR® Measuring Beakers with Handle, Porcelain
Catalog # 213-0408
Supplier:  VWR Collection
Supplier #:
undefined
VWR® Measuring Beakers with Handle, Porcelain
Catalog # 213-0408
Supplier:  VWR Collection
Supplier #:
Log in for availability
Select an Option
50 ml
Select an Option
01
Select an Option
1
About this item
Porcelain, glazed white.
  • Exceptional chemical resistance
  • Excellent thermal resistance
  • Robust, heavy construction
Cylindrical, with spout. Very easy to read, internal graduations.
Frequently Bought Together