Orders LinkContactUsLinkComponent
Column packing support, Quickfit®
Column packing support, Quickfit®
Supplier:   SciLabware
Column packing support, Quickfit®
Supplier:  SciLabware
Certificates
Column packing support, Quickfit®
Column packing support, Quickfit®
3 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
Designed to support Raschig rings within plain fractionating columns. Support is passed through socket of column and rests at the constriction above the cone.
Product Family Options
Product Information
  • Ø
    Cone size
    Availability
    Price
  • 16 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 56,40 €
    56,40 €
    1 * 5 ST
       56,40 €
  • 21 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,10 €
    20,10 €
    1 * 1 ST
       20,10 €
  • 29 mm
    34/35 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 84,00 €
    84,00 €
    1 * 5 ST
       84,00 €