Scheidetrichter, birnenförmig, Quickfit®
Supplier: SciLabware
Certificates
About this item
PYREX®, borosilicate glass.
- Without graduation
- Excellent chemical resistance
- Excellent temperature resistance
Funnel stem with cone to allow easy connection to Quickfit® flask with interchangeable PTFE key stopcock.
Certifications: Complies with ISO 4800 type 2.
Frequently Bought Together