ContactUsLinkComponent
Casting tape, Scotchcast™ Plus
Catalog # MMMEM82002A
Supplier:  3M MEDICAL
undefined
Casting tape, Scotchcast™ Plus
Catalog # MMMEM82002A
Supplier:  3M MEDICAL
Specifications
  • Description:
    Casting tape, black, 3,6 m roll, 50 mm width
  • Pk:
    10 Roll
Specifications
About this item
This casting tape consists of a knitted fiberglass fabric impregnated with polyurethane resin. Exposure to moisture or water initiates a chemical reaction which causes the tape to become rigid.
  • Finished casts produced from this casting tape are lightweight, strong, radiolucent, and porous
  • Scotchcast™ Plus casting tape contains a lubricating agent in the resin to provide a slippery feel when exposed to water