ContactUsLinkComponent
Silicone inserts for pipette containers
Silicone inserts for pipette containers
Catalog # SCUT3523213
Silicone inserts for pipette containers
Catalog # SCUT3523213
Specifications
  • Description:
    Silicone insert (2 mm thick), prevents damage to pipettes caused by sudden shock, for pipette containers VARICON RS (lower part) and SPECICON (lower and upper part)
  • Pk:
    1
Specifications
About this item
Silicone rubber for insertion in VARICON RS (lower part) and SPECICON (lower and upper part) pipette containers and it is designed to prevent damage to pipettes caused by sudden shock.
Thickness: 2 mm
Ø: 58 mm