CF™ dye streptavidin and monoclonal mouse anti-biotin conjugates
Supplier: Biotium
Certificates
About this item
CF™ dyes are superior to other fluorescent dyes for protein labelling due to advantages in brightness, photostability and water solubility.
Streptavidin conjugates are typically used as secondary reagents to detect biotinylated probes such as primary antibodies for flow cytometry, Western blotting, immunofluorescence staining and other applications.
Streptavidin conjugates are supplied in lyophilised solid format. After reconstitution with 0,5 ml PBS, streptavidin concentration is 2 mg/ml with 0,01% sodium azide. Monoclonal mouse anti-biotin antibody conjugates are supplied at 2 mg/ml in PBS pH 7,4 containing 50% glycerol, 2 mg/ml BSA (IgG-free and protease-free), and 0,05% sodium azide.
Specifications
- Host:
- Environmentally Preferable:
Specifications
Frequently Bought Together