Orders LinkContactUsLinkComponent
Disposable filters, Optimizer® DEV and DI
Disposable filters, Optimizer® DEV and DI
Supplier:   ENTEGRIS
Disposable filters, Optimizer® DEV and DI
Supplier:  ENTEGRIS
Certificates
Disposable filters, Optimizer® DEV and DI
Disposable filters, Optimizer® DEV and DI
45 Options Available Below
Product Details & Documents
These point-of-use filters meet small to mid-volume photochemical filtration needs for positive developer and DI water applications. Both Optimizer® DEV filters (for developer applications) and Optimizer® DI filters (for DI water applications) require no pre-wetting, so the filters function immediately after installation. The design also limits the handling of hazardous chemicals during the installation and promotes a healthier workplace. Do not use either filter with aromatic hydrocarbons.
  • Manufactured, tested, and packaged in a world-class, ISO 9001 Quality Systems Standard facility
  • O-ringless connections avoid potential sources of contamination
Membrane: Patented surface-modified hydrophilic ultra-high molecular weight polyethylene (UHMWPE); Supports, core and sleeve: HDPE; Disposable shell: PP
Maximum operating temperature: 60 °C
Maximum forward differential pressure: 2,8 bar at 25 °C
Maximum reverse differential pressure: 2,8 bar at 25 °C
Articles may not be available in all countries. Please contact your local Avantor office.
Product Family Options
Product Information
  • Type
    Pore size
    Filtration area
    Availability
    Price
  • DEV disposable filter with 1/4" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×97 mm
    0,03 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV disposable filter with 6 mm connection, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×122 mm
    0,03 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV disposable filter with 1/4" connection, 1/4" vent/drain compression seal fittings, Ø×L: 71×97 mm
    0,05 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV disposable filter with 1/4" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×97 mm
    0,05 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV disposable filter with 6 mm connection, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×122 mm
    0,05 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV disposable filter with 8 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×127 mm
    0,05 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV disposable filter with 1/4" connection, 1/4" vent/drain compression seal fittings, Ø×L: 71×97 mm
    0,1 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV disposable filter with 1/4" connection, 1/4" vent/drain flaretek fittings, Ø×L: 71×97 mm
    0,1 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV disposable filter with 1/4" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×97 mm
    0,1 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV disposable filter with 6 mm connection, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×122 mm
    0,1 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV disposable filter with 8 mm connection, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×127 mm
    0,1 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV-L disposable filter with 3/8" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×209 mm
    0,03 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV-L disposable filter with 1/2" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×175 mm
    0,05 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV-L disposable filter with 1/4" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×175 mm
    0,05 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV-L disposable filter with 3/8" connection, 1/4" vent/drain compression seal fittings, Ø×L: 71×209 mm
    0,05 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV-L disposable filter with 3/8" connection, 1/4" vent/drain Flowell™ fittings, Ø×L: 71×209 mm
    0,05 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV-L disposable filter with 3/8" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×209 mm
    0,05 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV-L disposable filter with 1/2" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×175 mm
    0,1 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV-L disposable filter with 1/4" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×175 mm
    0,1 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV-L disposable filter with 3/8" connection, 1/4" vent/drain compression seal fittings, Ø×L: 71×209 mm
    0,1 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DEV-L disposable filter with 3/8" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×209 mm
    0,1 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 1/4" connections, 1/4" vent/drain compression seal fittings, Ø×L: 71×97 mm
    0,05 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 1/4" connections, 1/4" vent/drain flaretek fittings, Ø×L: 71×97 mm
    0,05 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 1/4" connections, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×97 mm
    0,05 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 6 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×122 mm
    0,05 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI-L disposable filter with 3/8" connections, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×209 mm
    0,05 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI-L disposable filter with 8 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×205 mm
    0,05 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 1/4" connections, 1/4" vent/drain compression seal fittings, Ø×L: 71×97 mm
    0,1 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 1/4" connections, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×97 mm
    0,1 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI-L disposable filter with 3/8" connections, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×209 mm
    0,1 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI-L disposable filter with 8 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×205 mm
    0,1 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 1/4" connections, 1/4" vent/drain compression seal fittings, Ø×L: 71×97 mm
    0,2 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 1/4" connections, 1/4" vent/drain flaretek fittings, Ø×L: 71×97 mm
    0,2 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 1/4" connections, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×97 mm
    0,2 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 8 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×127 mm
    0,05 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI-L disposable filter with 1/2" connections, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×175 mm
    0,05 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI-L disposable filter with 1/4" connections, 1/4" vent/drain compression seal fittings, Ø×L: 71×175 mm
    0,05 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI-L disposable filter with 3/8" connections, 1/4" vent/drain flaretek fittings, Ø×L: 71×209 mm
    0,05 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI-L disposable filter with 3/8" connections, 1/4" vent/drain Flowell™ fittings, Ø×L: 71×209 mm
    0,05 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 6 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×122 mm
    0,1 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 8 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×127 mm
    0,1 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI-L disposable filter with 1/2" connections, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×175 mm
    0,1 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI disposable filter with 8 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×127 mm
    0,2 µm
    1800 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI-L disposable filter with 3/8" connections, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×209 mm
    0,2 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • DI-L disposable filter with 8 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×205 mm
    0,2 µm
    3600 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST