ContactUsLinkComponent
VWR®, Bottles, Wide Neck, with Screw Cap
VWR®, Bottles, Wide Neck, with Screw Cap
Catalog # 331-0205
Supplier:  VWR Collection
undefined
VWR®, Bottles, Wide Neck, with Screw Cap
Catalog # 331-0205
Supplier:  VWR Collection
Specifications
  • Capacity:
    285 ml
  • Environmentally Preferable:
  • Pk:
    100
Specifications
About this item
PE, translucent with black PP caps.
  • Good chemical resistance
  • Good thermal resistance
  • Leakproof
Perfect for sample collection, storage and transport. Ideal for liquids, pastes and granular material. The extra wide mouth allows easy filling and cleaning.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.