Orders LinkContactUsLinkComponent
J.T.Baker®, SPE Glass Column Processors
Supplier:   Avantor
J.T.Baker®, SPE Glass Column Processors
Supplier:  Avantor
Certificates
J.T.Baker®, SPE Glass Column Processors
J.T.Baker®, SPE Glass Column Processors
2 Options Available Below
Product Details & Documents
These glass column processors are specially designed glass vacuum chambers that allow multiple samples to be run simultaneously. The glass chamber allows unobstructed viewing of samples at all times.
  • Run multiple samples simultaneously
  • Unobstructed viewing of samples
  • Specially-designed, glass vacuum chambers
The SPE-12G processor allows up to 12 samples to be run simultaneously and includes 12 Luer-type fittings, 12 individual flow control stopcocks, 12 polypropylene collection needles, a vacuum gauge/controller, and a rack set that accommodates a variety of glassware.
The SPE-24G processor allows up to 24 samples to be processed in 20 to 30 minutes and includes 24 Luer-type fittings, 24 individual flow control stopcocks, 24 polypropylene collection needles, a vacuum gauge/controller, and a rack set that accommodates a variety of glassware.
 : Both units are supplied with a vacuum manifold, a sample collection rack with three support posts, four adjustable height shelves with support clips, and an instruction manual.
Product Family Options
Product Information
  • Availability
    Price
  • 6998-00
    SPE-12G column processor
    SPE-12G column processor
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 214,52 €
    2 214,52 €
    1 * 1 ST
       2 214,52 €
  • 7208-00
    SPE-24G column processor
    SPE-24G column processor
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 666,03 €
    3 666,03 €
    1 * 1 ST
       3 666,03 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Luer connector, male, PP
    Luer connector, male, PP
    2121-20
    Log in for availability
    1 * 12 ST - 209,83 €
Product Information
  • Catalog #
    Availability
    Price
  • Luer connector, female, PP
    Luer connector, female, PP
    2120-02
    Log in for availability
    1 * 12 ST - 121,12 €
Product Information
  • Catalog #
    Availability
    Price
  • Needles, stainless steel
    Needles, stainless steel
    7292-00
    Log in for availability
    1 * 1 Pack. - 487,96 €
Product Information
  • Catalog #
    Availability
    Price
  • Glass vacuum chamber, for SPE-24G column processor
    Glass vacuum chamber, for SPE-24G column processor
    7423-00
    Log in for availability
    1 * 1 ST - 516,47 €
Product Information
  • Catalog #
    Availability
    Price
  • Gasket seals, neoprene, for SPE-24G column processor
    Gasket seals, neoprene, for SPE-24G column processor
    7435-00
    Log in for availability
    1 * 1 ST - 88,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Needles, PP
    Needles, PP
    7436-00
    Log in for availability
    1 * 12 ST - 128,56 €
Product Information
  • Catalog #
    Availability
    Price
  • Vacuum gauge/controller assembly
    Vacuum gauge/controller assembly
    7437-00
    Log in for availability
    1 * 1 ST - 418,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Luer stopcocks, stainless steel
    Luer stopcocks, stainless steel
    4505.
    Log in for availability
    1 * 12 ST - 657,76 €
Product Information
  • Catalog #
    Availability
    Price
  • Stopcocks, PP
    Stopcocks, PP
    7166-00
    Log in for availability
    1 * 12 ST - 135,53 €
Product Information
  • Catalog #
    Availability
    Price
  • Luer stopcocks, PTFE
    Luer stopcocks, PTFE
    7514-00
    Log in for availability
    1 * 12 ST - 735,39 €