ContactUsLinkComponent
Cuvette for LKB analyser
Cuvette for LKB analyser
Catalog # SARS68.752
Supplier:  SARSTEDT
undefined
Cuvette for LKB analyser
Catalog # SARS68.752
Supplier:  SARSTEDT
Specifications
  • Ø×H:
    12×51 mm
  • Volume:
    3,5 ml
  • Description:
    Round cuvettes
  • Material:
    PP
  • Pk:
    5.000
Specifications
About this item
Polypropylene round opaque cuvette for sample preparation with LKB analyser.