Tubing, metal
Metal tubing is integrated insulated and flexible with connectors and fittings. This flexible metal tubing can be bolted together with the circulator and the external system, provides anti-slipping. Triple insulated tubing is used for wide temperature ranges.
Frequently Bought Together