Hermetic jars, wide mouth
Supplier: LAMAPLAST
Certificates
About this item
PP, white.
- Self-sealing screw cap
- Cap must be ordered seperately
- Good chemical resistance, good temperature resistance
Wide universal containers for creams, semi solid substances, powders and granulates.
Screw caps
Product Information
Frequently Bought Together