Order Entry
Austria
Orders LinkContactUsLinkComponent
Snap cap for solvent vials

Snap cap for solvent vials

Snap cap, 5 ml and 10 ml solvent vials, PTFE/silicone
Catalog # HEWL8010-0423
Supplier:   Agilent
Snap cap for solvent vials
Catalog # HEWL8010-0423
Supplier:  Agilent
Snap cap for solvent vials

Snap cap, 5 ml and 10 ml solvent vials, PTFE/silicone

Price
165,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
Specifications
  • For:
    5 ml and 10 ml solvent vials
  • Septum:
    PTFE/silicone
  • Description:
    Snap cap
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.