ContactUsLinkComponent
Capillary tubing kits
Capillary tubing kits
Catalog # HEWL5067-4601
Supplier:  Agilent
undefined
Capillary tubing kits
Catalog # HEWL5067-4601
Supplier:  Agilent
Specifications
  • Description:
    Solvent selection tubing kit
  • Pk:
    1
Specifications
About this item
Kit includes all capillaries and fittings, as well as bulk PEEK capillaries and a capillary cutter to add maximum flexibility.
  • Solvent selection kit- flow rate upto 10 ml/min