ContactUsLinkComponent
Autosampler vial sleeve
Autosampler vial sleeve
Catalog # PERCN9307027
Supplier:  PerkinElmer
Autosampler vial sleeve
Catalog # PERCN9307027
Supplier:  PerkinElmer
Specifications
  • For:
    200 µl and 300 µl autosampler vials
  • Description:
    12×32 mm clear glass support sleeve, open base
  • Environmentally Preferable:
  • Pk:
    25
Specifications
About this item
This open base clear glass support sleeve is used to offer microsampling capability in a standard 2 ml vial carousel or robotic autosampler.
  • 12×32 mm support sleeve
  • Accepts 200 and 300 µl vials