Slide cabinet
Catalog # LEIH3808500E
Supplier: LEICA HISTOLOGY
Specifications
- No. of slides:6430
- Colour:Blue
- Description:Slide cabinet
- Material:Steel
- Environmentally Preferable:
- Pk:1
Specifications
About this item
The slide cabinet is a storage system that can hold up to 6430 standard microscope slides.
- Stackable
- 14 drawers
- Black base plinth or wheeled plinth
Ordering information: Other colours available in option.