Orders LinkContactUsLinkComponent
Precision density hydrometers, M100
Precision density hydrometers, M100
Supplier:   Ludwig Schneider
Precision density hydrometers, M100
Supplier:  Ludwig Schneider
Certificates
Precision density hydrometers, M100
Precision density hydrometers, M100
14 Options Available Below
Product Details & Documents
These precision density hydrometers without thermometers are suitable for calibration.
Error limit: ±0,002 g/cm³
Reference temperature: 20 °C
Certifications: DIN 12791
Ordering information: Surface tension in class M (medium) on request.
Packaging: Individually in a square plastic sleeve.
Product Family Options
Surface tension: Class L up to 1000 g/cm³
Product Information
  • Model
    Range
    Division
    Temperature Range
    Length
    Availability
    Price
  • M100-060
    0,600 – 0,700 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,10 €
    28,10 €
    1 * 1 ST
       28,10 €
  • M100-070
    0,700 – 0,800 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,10 €
    28,10 €
    1 * 1 ST
       28,10 €
  • M100-080
    0,800 – 0,900 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,00 €
    26,00 €
    1 * 1 ST
       26,00 €
  • M100-090
    0,900 – 1,000 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,00 €
    26,00 €
    1 * 1 ST
       26,00 €
Surface tension: Class H above 1000 g/cm³
Product Information
  • Model
    Range
    Division
    Temperature Range
    Length
    Availability
    Price
  • SCNL3192005
    Density hydrometers, type M 100-100
    Density hydrometers, type M 100-100
    M100-100
    1,000 – 1,100 g/cm³
    0,002 g/cm³
    0...+30 °C
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,10 €
    28,10 €
    1 * 1 ST
       28,10 €
  • SCNL3192006
    Density hydrometers, type M 100-110
    Density hydrometers, type M 100-110
    M100-110
    1,100 – 1,200 g/cm³
    0,002 g/cm³
    0...+30 °C
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,10 €
    28,10 €
    1 * 1 ST
       28,10 €
  • M100-120
    1,200 - 1,300 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,10 €
    28,10 €
    1 * 1 ST
       28,10 €
  • M100-130
    1,300 - 1,400 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,80 €
    30,80 €
    1 * 1 ST
       30,80 €
  • M100-140
    1,400 - 1,500 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,80 €
    30,80 €
    1 * 1 ST
       30,80 €
  • M100-150
    1,500 - 1,600 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,80 €
    30,80 €
    1 * 1 ST
       30,80 €
  • M100-160
    1,600 - 1,700 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,90 €
    33,90 €
    1 * 1 ST
       33,90 €
  • M100-170
    1,700 - 1,800 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,90 €
    33,90 €
    1 * 1 ST
       33,90 €
  • M100-180
    1,800 - 1,900 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,80 €
    37,80 €
    1 * 1 ST
       37,80 €
  • M100-190
    1,900 - 2,000 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,80 €
    37,80 €
    1 * 1 ST
       37,80 €