Order Entry
Austria
ContactUsLinkComponent
HPLC columns, LC-NH₂, SUPELCOSIL, Supelco®
HPLC columns, LC-NH₂, SUPELCOSIL, Supelco®
Catalog # 59132.
Supplier:  Merck
HPLC columns, LC-NH₂, SUPELCOSIL, Supelco®
Catalog # 59132.
Supplier:  Merck

Specifications

  • Ø int.:
    4,60 mm
  • Particle Size:
    5 µm
  • Length:
    250 mm
  • Pk:
    1

Specifications

About this item

The LC-NH2-NP is an amino phase dedicated to normal phase chromatography suitable for separation of wate insoluble analytes like fat soluble vitamins.

  • Show stable retention in normal phase separations
  • Less sensitive to small or varying amounts of water in the mobile phase, relative to unmodified silica
  • Provide excellent separations of fat-soluble vitamins