SPE Plates, 96-Well, Hypersep™ Hypercarb
Supplier: Thermo Fisher Scientific
Certificates
About this item
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Guanidinium hydrochloride 8 mol/l in aqueous solution
Supplier: Thermo Fisher Scientific
Screw caps for certified screw top vials
Supplier: Agilent
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection