Order Entry
Austria
ContactUsLinkComponent
di-Lithium tetraborate ≥99.0%, AnalaR NORMAPUR® ACS for X-ray analysis
Corrosive
Health
Irritant
di-Lithium tetraborate ≥99.0%, AnalaR NORMAPUR® ACS for X-ray analysis
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Health
Irritant
di-Lithium tetraborate ≥99.0%, AnalaR NORMAPUR® ACS for X-ray analysis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Bulk density (g/ml):
    Min. 0.25
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insoluble matter:
    Max. 100 ppm
  • Loss on fusion (950°C):
    Max. 2.0 %
  • Phosphorus compounds (as PO4):
    Max. 40 ppm
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Al (Aluminium):
    Max. 10 ppm
  • Ca (Calcium):
    Max. 30 ppm
  • Fe (Iron):
    Max. 10 ppm
  • K (Potassium):
    Max. 30 ppm
  • Mg (Magnesium):
    Max. 5 ppm
  • Na (Sodium):
    Max. 50 ppm
  • Si (Silicium):
    Max. 80 ppm
  • Conforms to ACS:
    Passes test

Specification Test Results

Frequently Bought Together