ContactUsLinkComponent
Dosing bottles, HDPE
Dosing bottles, HDPE
  216-5680
 :  NEOLAB
undefined
Dosing bottles, HDPE
  216-5680
 :  NEOLAB
  • Capacity:
    120 ml
  • Pk:
    1
HDPE, with metering device.
Suitable for the application which requires fine and pointedly dosing of many liquids, e.g. oils and petrol, especially for the wetting of cleaning wipes with alcohols. The feeding pipe even brings the last remains up. When not used, the flask is hermetical closed so that volatile substances cannot escape.