Order Entry
Austria
ContactUsLinkComponent
 
pH/mV/°C/ion meters, handheld, Seven2Go™ S2 / S8 Pro
 
undefined
pH/mV/°C/ion meters, handheld, Seven2Go™ S2 / S8 Pro

 

  • Catalog No:
  • 662-1838
  • 662-1844
  • 662-1845
  • 662-1846
  • 662-1847
  • 662-1835
  • 662-1848
  • 662-1834
  • 662-1837
  • 662-1836
  • Model:
  • S2
  • S8
  • S8
  • S8
  • S8
  • S2
  • S8
  • S2
  • S2
  • S2
  • pH range:
  • −2...+20
  • −2...+20
  • −2...+20
  • −2...+20
  • −2...+20
  • −2...+20
  • −2...+20
  • −2...+20
  • −2...+20
  • −2...+20
  • pH resolution:
  • 0,01
  • 0,001
  • 0,001
  • 0,001
  • 0,001
  • 0,01
  • 0,001
  • 0,01
  • 0,01
  • 0,01
  • pH accuracy:
  • ±0,01
  • ±0,001
  • ±0,001
  • ±0,001
  • ±0,001
  • ±0,01
  • ±0,001
  • ±0,01
  • ±0,01
  • ±0,01
  • pH calibration:
  • 5 points with 4 predefined or user-defined buffers
  • 5 points with 8 predefined or user-defined buffers
  • 5 points with 8 predefined or user-defined buffers
  • 5 points with 8 predefined or user-defined buffers
  • 5 points with 8 predefined or user-defined buffers
  • 5 points with 4 predefined or user-defined buffers
  • 5 points with 8 predefined or user-defined buffers
  • 5 points with 4 predefined or user-defined buffers
  • 5 points with 4 predefined or user-defined buffers
  • 5 points with 4 predefined or user-defined buffers
  • Automatic buffer recognition:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • mV range:
  • ±1999
  • ±2000
  • ±2000
  • ±2000
  • ±2000
  • ±1999
  • ±2000
  • ±1999
  • ±1999
  • ±1999
  • mV resolution:
  • 1
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • 1
  • 0,1
  • 1
  • 1
  • 1
  • mV accuracy:
  • ±1
  • ±0,1
  • ±0,1
  • ±0,1
  • ±0,1
  • ±1
  • ±0,1
  • ±1
  • ±1
  • ±1
  • ISE range:
  • -
  • 1,00e-09...9,99e09
  • 1,00e-09...9,99e09
  • 1,00e-09...9,99e09
  • 1,00e-09...9,99e09
  • -
  • 1,00e-09...9,99e09
  • -
  • -
  • -
  • ISE resolution:
  • -
  • 3 or 4 digits (auto)
  • 3 or 4 digits (auto)
  • 3 or 4 digits (auto)
  • 3 or 4 digits (auto)
  • -
  • 3 or 4 digits (auto)
  • -
  • -
  • -
  • ISE accuracy:
  • -
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • -
  • ±0,5%
  • -
  • -
  • -
  • ISE calibration:
  • -
  • 5 points, 1 user-defined ion-standard
  • 5 points, 1 user-defined ion-standard
  • 5 points, 1 user-defined ion-standard
  • 5 points, 1 user-defined ion-standard
  • -
  • 5 points, 1 user-defined ion-standard
  • -
  • -
  • -
  • ISE units:
  • -
  • mg/l, mmol/l, mol/l, %, ppm, pX
  • mg/l, mmol/l, mol/l, %, ppm, pX
  • mg/l, mmol/l, mol/l, %, ppm, pX
  • mg/l, mmol/l, mol/l, %, ppm, pX
  • -
  • mg/l, mmol/l, mol/l, %, ppm, pX
  • -
  • -
  • -
  • Temperature range:
  • −5...+105 °C
  • −5...+130 °C (ATC), −30...+130 °C (MTC)
  • −5...+130 °C (ATC), −30...+130 °C (MTC)
  • −5...+130 °C (ATC), −30...+130 °C (MTC)
  • −5...+130 °C (ATC), −30...+130 °C (MTC)
  • −5...+105 °C
  • −5...+130 °C (ATC), −30...+130 °C (MTC)
  • −5...+105 °C
  • −5...+105 °C
  • −5...+105 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,5 °C
  • ±0,2 °C; (±0,5 °C if T<0 °C or T>105 °C)
  • ±0,2 °C; (±0,5 °C if T<0 °C or T>105 °C)
  • ±0,2 °C; (±0,5 °c if T<0 °C or T>105 °C)
  • ±0,2 °C; (±0,5 °c if T<0 °C or T>105 °C)
  • ±0,5 °C
  • ±0,2 °C; (±0,5 °C if T<0 °C or T>105 °C)
  • ±0,5 °C
  • ±0,5 °C
  • ±0,5 °C
  • Temperature compensation:
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Display:
  • Segmented LCD
  • Graphic LCD
  • Graphic LCD
  • Graphic LCD
  • Graphic LCD
  • Segmented LCD
  • Graphic LCD
  • Segmented LCD
  • Segmented LCD
  • Segmented LCD
  • Power supply:
  • 4× 1,5 V AA batteries or 1,3 V rechargeable battery with NiMH cells
  • 4× 1,5 V AA batteries or 1,3 V rechargeable battery with NiMH cells
  • 4× 1,5 V AA batteries or 1,3 V rechargeable battery with NiMH cells
  • 4× 1,5 V AA batteries or 1,3 V rechargeable battery with NiMH cells
  • 4× 1,5 V AA batteries or 1,3 V rechargeable battery with NiMH cells
  • 4× 1,5 V AA batteries or 1,3 V rechargeable battery with NiMH cells
  • 4× 1,5 V AA batteries or 1,3 V rechargeable battery with NiMH cells
  • 4× 1,5 V AA batteries or 1,3 V rechargeable battery with NiMH cells
  • 4× 1,5 V AA batteries or 1,3 V rechargeable battery with NiMH cells
  • 4× 1,5 V AA batteries or 1,3 V rechargeable battery with NiMH cells
  • Compliance:
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • Data logging capacity:
  • 200 values
  • 2000 values
  • 2000 values
  • 2000 values
  • 2000 values
  • 200 values
  • 2000 values
  • 200 values
  • 200 values
  • 200 values
  • Connections:
  • BNC (1012 Ω) / RCA (cinch)
  • BNC (1012 Ω) / RCA (cinch) / 2 mm banana
  • BNC (1012 Ω) / RCA (cinch) / 2 mm banana
  • BNC (1012 Ω) / RCA (cinch) / 2 mm banana
  • BNC (1012 Ω) / RCA (cinch) / 2 mm banana
  • BNC (1012 Ω) / RCA (cinch)
  • BNC (1012 Ω) / RCA (cinch) / 2 mm banana
  • BNC (1012 Ω) / RCA (cinch)
  • BNC (1012 Ω) / RCA (cinch)
  • BNC (1012 Ω) / RCA (cinch)
  • Outputs:
  • Micro USB
  • Micro USB
  • Micro USB
  • Micro USB
  • Micro USB
  • W×D×H:
  • 220×70×35 mm
  • 220×70×35 mm
  • 220×70×35 mm
  • 220×70×35 mm
  • 220×70×35 mm
  • 220×70×35 mm
  • 220×70×35 mm
  • 220×70×35 mm
  • 220×70×35 mm
  • 220×70×35 mm
  • Weight:
  • 0,270 kg
  • 0,290 kg
  • 0,290 kg
  • 0,290 kg
  • 0,290 kg
  • 0,270 kg
  • 0,290 kg
  • 0,270 kg
  • 0,270 kg
  • 0,270 kg

 

Frequently Bought Together