Accessories for SPE columns
Supplier: Merck
Certificates
About this item
Frequently Bought Together
VWR®, Test Tubes Racks
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Pig Pancreatin, Protease, Lipase, Amylase (from Pancreas)
Supplier: MERCK PRODUCTION CHEMICALS
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Accessories for vortex mixers, MS 3
Supplier: IKA
Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Tubing clip
Supplier: VICI JOUR RESEARCH
Empty solid phase extraction cartridges, Bond Elut™
Supplier: VARIAN
GC ferrules, SilTite™ FingerTite
Supplier: Trajan Scientific and Medical