ContactUsLinkComponent
Detergent, Trump Supra plus, Ecolab®
Detergent, Trump Supra plus, Ecolab®
Catalog # KAMM9013840
Supplier:  KAMMERER
Detergent, Trump Supra plus, Ecolab®
Catalog # KAMM9013840
Supplier:  KAMMERER
Specifications
  • Description:
    Trump Supra plus, concentrated alkaline detergent
  • Packed:
    1 canister of 12 kg
  • Pk:
    12 kg
Specifications
About this item
Trump Supra plus is a highly concentrated alkaline detergent for use in dishwashing/glassware washing machines.
  • Active chlorine for hygienically clean finish
  • Powerful cleaning action even at high water hardness
Compatible with materials that are alkaline and bleach resistant - not suitable for aluminium