Alcohol meters
Supplier: GERING
Certificates
About this item
Alcohol meters with a standard temperature of 20 °C.
Certifications: DIN 12803
Ordering information: Includes a thermometer.
Frequently Bought Together
Alcohol meters with a standard temperature of 20 °C.