Order Entry
Austria
ContactUsLinkComponent
RNA isolation, total RNA kit, E.Z.N.A.® MicroElute
RNA isolation, total RNA kit, E.Z.N.A.® MicroElute
Supplier:  OMEGA BIO-TEK
Certificates
RNA isolation, total RNA kit, E.Z.N.A.® MicroElute
Supplier:  OMEGA BIO-TEK
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Micro
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    FFPE tissues
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Sample size:
    5×105 cells or 5 mg tissue
  • Description:
    E.Z.N.A.® MicroElute® Total RNA Kit

Specifications

Related Information

Frequently Bought Together