Order Entry
Austria
ContactUsLinkComponent
E.Z.N.A.® Plasmid Mini kit II, Q-spin (without lid)
E.Z.N.A.® Plasmid Mini kit II, Q-spin (without lid)
Supplier:  OMEGA BIO-TEK
Certificates
undefined
E.Z.N.A.® Plasmid Mini kit II, Q-spin (without lid)
Supplier:  OMEGA BIO-TEK
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini plus
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Sample size:
    3 - 15 ml
  • Description:
    E.Z.N.A.® Plasmid Mini Kit II (Q-Spin column)

Specifications

Related Information

Frequently Bought Together