Orders LinkContactUsLinkComponent
Dust mop frame
Dust mop frame
Dust mop frame, 600 mm
Catalog # 129-1077
Supplier:   FHP VILEDA SCS
Dust mop frame
Catalog # 129-1077
Supplier:  FHP VILEDA SCS
Dust mop frame
Dust mop frame
Dust mop frame, 600 mm
Price
46,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Flexible dust mop frame developed to complement the range of dust mop cloths.
  • Developed to complement the range of dust mop cloths
  • Long-lasting, durable rubber blades ensure maximum surface contact
  • Universal connector fits all handles
Long-lasting, durable rubber blades ensure maximum surface contact and the universal connector fits all handles. The durable and flexible design allows the frame to move smoothly over even or uneven floors.
Specifications
  • Description:
    Dust mop frame
  • Length:
    600 mm