Orders LinkContactUsLinkComponent
Sample tubes
Sample tubes
Supplier:   METROHM
Sample tubes
Supplier:  METROHM
Certificates
2 Options Available Below
Product Details & Documents
Ideal for IC sample processors and VA autosamplers.
Product Family Options
Product Information
  • Capacity
    Length
    Ext.Ø
    Material
    Availability
    Price
  • 11 ml
    108 mm
    16 mm
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 1 900,00 €
    1 900,00 €
    1 * 2.000 ST
       1 900,00 €
  • 11 ml
    108 mm
    16 mm
    PP
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 355,00 €
    355,00 €
    1 * 200 ST
       355,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Perforated stopper, PP, for 2,5 and 11 ml sample tubes
    Perforated stopper, PP, for 2,5 and 11 ml sample tubes
    METR6.2743.077
    Log in for availability
    1 * 200 ST - 107,00 €