Order Entry
Austria
ContactUsLinkComponent
Mag-Bind® Total RNA 96 kit
Mag-Bind® Total RNA 96 kit
Supplier:  OMEGA BIO-TEK
Certificates
Mag-Bind® Total RNA 96 kit
Supplier:  OMEGA BIO-TEK

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA

Specifications

Related Information

Frequently Bought Together