Order Entry
Austria
Orders LinkContactUsLinkComponent
 

E.Z.N.A.® Plasmid Maxi Kit

E.Z.N.A.® Plasmid Maxi Kit
 
E.Z.N.A.® Plasmid Maxi Kit
E.Z.N.A.® Plasmid Maxi Kit

 

 

The E.Z.N.A.® Plasmid Maxi Kit is the maxi scale system to deliver high-quality plasmid DNA using the spin column format, without costly accessories. The HiBind® DNA Maxi column facilitates the binding, washing, and elution steps, thus enabling multiple samples to be simultaneously processed. Although yields vary according to plasmid copy number, E.coli strain, and conditions of growth, 50 to 200 ml of overnight culture in LB medium typically produces 0,5 to 1 mg of high copy number plasmid DNA. Up to 500 ml of culture may be processed when working with low copy number plasmid. The product is suitable for automated fluorescent DNA sequencing (typical reads exceed 800 bp), restriction enzyme digestion, transfection of mammalian cells, transcription invitro, and other applications.

  • Purification of plasmid DNA in < 60 minutes
  • No phenol/chloroform extractions
  • Yield up to 1 mg of Plasmid DNA
  • DNA suitable for downstream applications
 

 

 

  • Catalog No:
  • D6922-02
  • D6922-04
  • D6922-00
  • Description:
  • E.Z.N.A.® Plasmid Maxi Kit I
  • E.Z.N.A.® Plasmid Maxi Kit I
  • E.Z.N.A.® Plasmid Maxi Kit I
  • Target molecule:
  • Plasmid
  • Plasmid
  • Plasmid
  • Refine target molecule:
  • Plasmid
  • Plasmid
  • Plasmid
  • Sample type:
  • Bacteria
  • Bacteria
  • Bacteria
  • Method / Format:
  • Column (silica-based)
  • Column (silica-based)
  • Column (silica-based)
  • Prep size:
  • Maxi
  • Maxi
  • Maxi
  • Sample size:
  • 50 - 200 ml
  • 50 - 200 ml
  • 50 - 200 ml

 

Product Family Options

 

  • Description
    Pk
     
     
  • E.Z.N.A.® Plasmid Maxi Kit I
    20 Tests
       
       
    1 * 20 Tests
    1 * 20 Tests - 416,00 €
    416,00 €
    1 * 20 Tests
       416,00 €

     
     
    Specifications
    Cat. No.
    D6922-02
    Description
    E.Z.N.A.® Plasmid Maxi Kit I
    Target molecule
    Plasmid
    Refine target molecule
    Plasmid
    Sample type
    Bacteria
    Method / Format
    Column (silica-based)
    Prep size
    Maxi
    Sample size
    50 - 200 ml
  • E.Z.N.A.® Plasmid Maxi Kit I
    100 Tests
       
       
    1 * 100 Tests
    1 * 100 Tests - 1 310,00 €
    1 310,00 €
    1 * 100 Tests
       1 310,00 €

     
     
    Specifications
    Cat. No.
    D6922-04
    Description
    E.Z.N.A.® Plasmid Maxi Kit I
    Target molecule
    Plasmid
    Refine target molecule
    Plasmid
    Sample type
    Bacteria
    Method / Format
    Column (silica-based)
    Prep size
    Maxi
    Sample size
    50 - 200 ml
  • E.Z.N.A.® Plasmid Maxi Kit I
    2 Tests
       
       
    1 * 2 Tests
    1 * 2 Tests - 66,10 €
    66,10 €
    1 * 2 Tests
       66,10 €

     
     
    Specifications
    Cat. No.
    D6922-00
    Description
    E.Z.N.A.® Plasmid Maxi Kit I
    Target molecule
    Plasmid
    Refine target molecule
    Plasmid
    Sample type
    Bacteria
    Method / Format
    Column (silica-based)
    Prep size
    Maxi
    Sample size
    50 - 200 ml