ContactUsLinkComponent
Handheld digital microscope, Dino-Lite Medical, IriScope
Handheld digital microscope, Dino-Lite Medical, IriScope
Catalog # 630-2615
undefined
Handheld digital microscope, Dino-Lite Medical, IriScope
Catalog # 630-2615
Specifications
  • Magnification:
    10 - 20×
  • Model:
    IriScope
  • Resolution:
    1,3 MP
  • Description:
    Handheld microscope for iris examinations
  • Seq:
    01
  • Illumination:
    8 LEDs (white + yellow)
  • Pk:
    1
Specifications
About this item
The IriScope enables users to make razor-sharp images of the iris.
  • Handy, easy to use
  • Magnification of 10 to 20 times allows users to see details that can hardly be seen by the naked eye
  • Specially designed front cap can be placed on the eye socket to get the right distance for a perfect picture
  • Built-in LEDs (white + yellow) provide better images of dark irises
  • High speed USB 2.0 output
Certifications: Medical Device Class 1 – according to Directive 93/42/EEC modified 2007/47/EC.
Delivery: Supplied with protecting pouch, software CD and calibration target.