Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

45 l, 600×400×236 mm
Catalog # SEMA2716
Supplier:   SEMADENI
Storage Boxes
Catalog # SEMA2716
Supplier:  SEMADENI
Storage Boxes

45 l, 600×400×236 mm

Price
75,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, grey.

  • Stackable
  • Closed bottom
  • Perforated walls

The standard light weight containers allows visual inspection of the contents.

Specifications
  • Capacity:
    45 l
  • W×D×H:
    600×400×236 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.