String mops, NovaKnit™
About this item
A series of robust and absorbent tubular knit string mops for cleaning and liquid pick-up in controlled and critical environments. Available with a choice of durable, reusable fabrics.
- Robust, tubular knit floor mop
- Ribbed strands increase absorbency
- Looped design with no exposed edges to ensure low particulates
- More absorbent and durable than traditional cleanroom mops
- Use with MH-1 handle or QDMH-1 MicroMop adapter with SSU series handles
- Available gamma irradiated for aseptic environments
- Holds up to strong disinfectants
- Launderable for reuse
- Autoclavable
The Micronova NovaKnit™ String Mops feature unique ribbing and a looped strand design to produce a mop that is both low-linting and highly absorbent. Available in three materials to match cleaning needs: 100% Microdenier polyester, for fine cleaning and absorbency, 100% polyester for a smooth clean, and a 75% polyester/25% nylon blend for applications requiring more absorbency.
Weight: 12 oz. (340 g)
Ordering information: All components must be ordered separately.
4 Options Available Below
Product Details & Documents
A series of robust and absorbent tubular knit string mops for cleaning and liquid pick-up in controlled and critical environments. Available with a choice of durable, reusable fabrics.
- Robust, tubular knit floor mop
- Ribbed strands increase absorbency
- Looped design with no exposed edges to ensure low particulates
- More absorbent and durable than traditional cleanroom mops
- Use with MH-1 handle or QDMH-1 MicroMop adapter with SSU series handles
- Available gamma irradiated for aseptic environments
- Holds up to strong disinfectants
- Launderable for reuse
- Autoclavable
The Micronova NovaKnit™ String Mops feature unique ribbing and a looped strand design to produce a mop that is both low-linting and highly absorbent. Available in three materials to match cleaning needs: 100% Microdenier polyester, for fine cleaning and absorbency, 100% polyester for a smooth clean, and a 75% polyester/25% nylon blend for applications requiring more absorbency.
Weight: 12 oz. (340 g)
Ordering information: All components must be ordered separately.
Documents
TDS NovaKnit™ String Mops
Datasheet
Specifications for Product Family
Specifications
- Catalog No:
- MICRMP5231-12WIR
- MICRMP52-31-12W
- MICRVW51-31-12W
- MICRMD5131-12WIR
- Environmentally Preferable:
- Waste Reducing
Specifications
Product Family Options
Mop Heads
Product Information
- MaterialAvailabilityPrice
- MICRMP5231-12WIRPolyester looped string mop, irradiatedSpecifications:
More
Specifications
Cat. No.MICRMP5231-12WIREnvironmentally Preferable - MICRMP52-31-12WPolyester looped string mopSpecifications:
More
Specifications
Cat. No.MICRMP52-31-12WEnvironmentally Preferable - MICRVW51-31-12WMicroDenier polyester looped string mop, irradiatedMicroDenier polyester looped string mop, irradiatedPolyesterLog in for availability1 * 1 ST34,60 €Specifications:
Waste Reducing More
Specifications
Cat. No.MICRVW51-31-12WEnvironmentally PreferableWaste ReducingWaste Reducing
Product Information
- MaterialAvailabilityPrice
- MICRMD5131-12WIRMops, NovaKnit™, microfiber, irradiatedSpecifications:
More
Specifications
Cat. No.MICRMD5131-12WIREnvironmentally Preferable
Adapters and Handles
Product Information
- Catalog #DescriptionAvailabilityPrice
- MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handlesLog in for availability1 * 1 ST157,00 €
Accessories
Product Information
- Catalog #LengthMaterialAvailabilityPrice
- Stainless steel quick disconnect handleStainless steel quick disconnect handle129-0062155 cmFibreglassLog in for availability1 * 1 ST - 165,00 €