Orders LinkContactUsLinkComponent
Microfiber string mops, M-Zone MegaSorb™
Microfiber string mops, M-Zone MegaSorb™
   Sustainable Options Available
Supplier:   MICRONOVA
Microfiber string mops, M-Zone MegaSorb™
Supplier:  MICRONOVA
   Sustainable Options Available
Certificates
Microfiber string mops, M-Zone MegaSorb™
3 Options Available Below
Product Details & Documents
A highly absorbent mop that combines the cleaning efficiency of microfibre with the ease of a looped string mop. The tufted strands are comprised of polyester and nylon filaments, designed to pick up liquids and tiny particles in controlled and critical environments.
  • Highly absorbent and low-linting
  • Polyester and nylon tufted microfibre tubes
  • Picks up tiny particles where cotton fibres just roll over
  • White microfibre option includes an additional scrubbing band
  • Use with MH-1 handle or QDMH-1 MicroMop adapter with SSU series handles
  • Compatible with alcohol and strong disinfectants
  • Launderable for reuse
  • Autoclavable
The M-Zone™ MegaSorb™ Microfibre String Mop features distinctive microfiber tubes with tufted strands made of polyester and nylon filaments, 30 times smaller than cotton fibres, that work to pick up tiny particles.
Specifications for Product Family
Specifications
  • Catalog No:
  • MICRJZMZCRMS-10
  • MICRMZCRMS-10IR
  • 129-0619
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
Specifications
Product Family Options
Mop heads
Product Information
  • Colour
    Material
    Availability
    Price
  • MICRJZMZCRMS-10
    MegaSorb™ mop head, 10 oz., bulk pack
    MegaSorb™ mop head, 10 oz., bulk pack
    Blue
    Microfibre
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,90 €
    36,90 €
    1 * 1 ST
       36,90 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRJZMZCRMS-10
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • MICRMZCRMS-10IR
    MegaSorb™ mop head, 10 oz., irradiated
    MegaSorb™ mop head, 10 oz., irradiated
    White
    Microcon blue vinyl
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 231,00 €
    231,00 €
    1 * 12 ST
       231,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MICRMZCRMS-10IR
    Environmentally Preferable
  • 129-0619
    MegaSorb™ mop head, 12 oz.
    MegaSorb™ mop head, 12 oz.
    White
    Microfibre
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 583,00 €
    583,00 €
    1 * 12 ST
       583,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    129-0619
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
Accessories
Product Information
  • Catalog #
    Length
    Availability
    Price
  • MicroMop® handle, Fibreglass
    MicroMop® handle, Fibreglass
    MICRMHC-1
    1524 mm
    Log in for availability
    1 * 1 ST - 72,20 €
Product Information
  • Catalog #
    Availability
    Price
  • MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    129-0170
    Log in for availability
    1 * 1 ST - 157,00 €
Product Information
  • Catalog #
    Length
    Material
    Availability
    Price
  • Stainless steel quick disconnect extendable handle
    Stainless steel quick disconnect extendable handle
    129-0551
    965 - 1829 mm
    Stainless steel 304
    Log in for availability
    1 * 1 ST - 358,00 €
Product Information
  • Catalog #
    Length
    Availability
    Price
  • MicroMop® handle, Fibreglass/PP
    MicroMop® handle, Fibreglass/PP
    129-0165
    1626 mm
    Log in for availability
    1 * 1 ST - 85,50 €