ContactUsLinkComponent
Disinfectants for use in Ultrasonic baths, STAMMOPUR DR 8
Disinfectants for use in Ultrasonic baths, STAMMOPUR DR 8
Catalog # BAND972
undefined
Disinfectants for use in Ultrasonic baths, STAMMOPUR DR 8
Catalog # BAND972
Specifications
  • Description:
    STAMMOPUR DR 8, disinfectant and cleaning concentrate
  • Pk:
    2 L
  • For:
    SUPER, DIGITEC, DIGITAL, LONGLIFE
Specifications
About this item
STAMMOPUR DR 8 concentrate is designed for cleaning of highly sensitive instruments using short sonication times in ultrasonic baths.
  • Rapid disinfection and efficient cleaning in ultrasonic baths
  • Formulation is free of aldehudes, phenols and chlorine
  • Broad range spectrum of activity- bactericidal, tuberculocidal, yeasticidal
  • Virucidal against BVDV, Vaccinia, Papova, HBV, HCV, HIV, A-H5N1
  • High material compatibility: Metal, glass, porcelain, plastics and rubber