Orders LinkContactUsLinkComponent
Forceps
 :   ENTEGRIS
Forceps
 :  ENTEGRIS
Forceps
ETFE, white. Durable forceps for substrate handling.
  • Different tip styles available
  • Packaged in protective cases
Product Family Options
  • Type
    Length
  • Straight, 0,3 mm thick tip
    124 mm
    1 * 4 ST
    1 * 4 ST
  • Straight, rounded tip (1,5 mm radius, 3,0 mm thickness)
    124 mm
    1 * 4 ST
    1 * 4 ST
  • Straight, rounded tip (0,8 mm radius)
    127 mm
    1 * 4 ST
    1 * 4 ST
  • Curved, tapered thin tip
    127 mm
    1 * 4 ST
    1 * 4 ST
  • Straight, rounded tip (1,5 mm radius, 3,0 mm thickness)
    178 mm
    1 * 4 ST
    1 * 4 ST