Safety spectacles, Virtua™ AP
Supplier: 3M
Jump to:
About this item
Lightweight, stylish designed safety glasses Virtua™ AP protective eyewear features a streamline wrap-around design for excellent coverage and good field of vision.
- Integrated brow bar for added protection
- Lightweight for maximum comfort
- Strong polycarbonate lenses featuring anti-scratch coating for improved vision and extended durability
Weight: 26 g
: EN 166:2001
2 Options Available Below
Product Details & Documents
Lightweight, stylish designed safety glasses Virtua™ AP protective eyewear features a streamline wrap-around design for excellent coverage and good field of vision.
- Integrated brow bar for added protection
- Lightweight for maximum comfort
- Strong polycarbonate lenses featuring anti-scratch coating for improved vision and extended durability
Weight: 26 g
: EN 166:2001
Product Family Options
Product Information