Order Entry
Austria
ContactUsLinkComponent
VWR®, Strontium 1000 mg/L in water standard for ion chromatography
VWR®, Strontium 1000 mg/L in water standard for ion chromatography
Catalog # 84994.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Strontium 1000 mg/L in water standard for ion chromatography
Catalog # 84994.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Strontium
  • Symbol:
    Sr

Specifications

About this item

Certifications: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.