VWR®, Glycolate 1,000 mg/L in water standard for ion chromatography
Artikelnr. 84968.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:1000 mg/l
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Description:Glycolate standard solution
Specifications
About this item
Certifications: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.