Sling psychrometer
Catalog # SIMETM0081
Supplier: Industrial Physics
Specifications
- Description:Sling psychrometer
- Pk:1
Specifications
About this item
The psychrometer is designed to measure the percent relative humidity on the basis of the wet bulb-dry bulb thermometer principle.
- Conversion scale
- Plastic case on the instrument
- Relative humidity percent calculations