ContactUsLinkComponent
Ultrapure water systems, Barnstead™ MicroPure™
Ultrapure water systems, Barnstead™ MicroPure™
  • Catalog No:
  • BARN50132374
  • BARN50132373
  • BARN50132372
  • BARN50132370
  • Model:
  • UV, with tank
  • UV
  • UV/UF, with tank
  • UV/UF
  • Feed water:
  • RO, DI or distilled water
  • RO, DI or distilled water
  • RO, DI or distilled water
  • RO, DI or distilled water
  • Flow rate:
  • 1,0 l/min
  • 1,5 l/min
  • 0,8 l/min
  • 1,0 l/min
  • Conductivity:
  • 0,055 µS/cm
  • 0,055 µS/cm
  • 0,055 µS/cm
  • 0,055 µS/cm
  • TOC content:
  • 1 - 5 ppb
  • 1 - 5 ppb
  • 1 - 5 ppb
  • 1 - 5 ppb
  • Resistivity at +25 °C:
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • Bacteria:
  • <1 CFU/ml
  • <1 CFU/ml
  • <1 CFU/ml
  • <1 CFU/ml
  • W×D×H:
  • 305×390×545 mm
  • 305×300×545 mm
  • 305×390×545 mm
  • 305×300×545 mm
  • Weight:
  • 17 kg
  • 15 kg
  • 17 kg
  • 15 kg
Frequently Bought Together