Order Entry
Austria
ContactUsLinkComponent
Self-adhesive labels, DURAN™ YOUTILITY
Self-adhesive labels, DURAN™ YOUTILITY
Catalog # SCOT294010203
Supplier:  DWK Life Sciences
undefined
Self-adhesive labels, DURAN™ YOUTILITY
Catalog # SCOT294010203
Supplier:  DWK Life Sciences

Specifications

  • Colour:
    White
  • Description:
    YOUTILITY labels, 100, on sheets
  • Dimensions:
    36×70 mm
  • Material:
    Polyester
  • Pk:
    1

Specifications

About this item

These self-adhesive, white polyester labels are designed for use with the DURAN™ YOUTILITY bottle system.

  • Chemical resistant
  • Temperature resistant at −40 to +150 °C
  • Non-toxic, and lead free
  • Autoclave/steam sterilisation Class 1 process indicator (ANSI/AAMI/ISO 11140-1:2005)

Surface will accept a variety of inks, including copier, printer, ballpoint and technical markers.