Orders LinkContactUsLinkComponent
Digital clock/timer
Digital clock/timer
Digital clock/timer, capacity of 24 h, White/grey
Catalog # COMAUTL882
Supplier:   COMARK
Digital clock/timer
Catalog # COMAUTL882
Supplier:  COMARK
Digital clock/timer
Digital clock/timer, capacity of 24 h, White/grey
Price
106,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This kitchen clock has a countdown capacity of 24 hours. All of its three functions − clock, timer and countdown − can operate simultaneously.
  • Built-in memory allows storage of a pre-set time
  • Count up in 1 second resolution up to 24 hours
Certifications: Compliant with HACCP (hazard analysis and critical control points) procedures.
Delivery information: Incl. magnet, pocket clip, stand and battery.
Specifications
  • Colour:
    White/grey
  • Description:
    Digital clock/timer, capacity of 24 h