Orders LinkContactUsLinkComponent
Baths and circulators, polycarbonate
Supplier:   HUBER
Baths and circulators, polycarbonate
Supplier:  HUBER
Certificates
Baths and circulators, polycarbonate
6 Options Available Below
Product Details & Documents
Heating circulation bath thermostats with polycarbonate or stainless steel baths, which are simple to use and are suitable for a variety of applications such as: sample temperature control, analysis, materials testing or checking food. The units offer high temperature stability and are fitted with over-temperature and low level protection suitable for Safety Class III/FL (DIN 12876) for use with flammable fluids. A powerful pressure/suction pump ensures optimum circulation and temperature homogenity in the bath and also allows temperature control of externally attached applications using a pump adapter (accessory). There is a range of accessories including inserts, variable adjustable platforms, bath covers, probes as well as hoses, temperature control fluids and various adapters.
  • Pilot ONE® controller with Plug and Play technology, 14,5 cm touchscreen and comfortable menu navigation
  • Precise temperature control
  • Overtemperature and low level protection
  • Powerful pressure/suction pump
  • Programmer with calendar and clock functions, allows 5-point calibration
  • PT100 sensor connection, USB and LAN interfaces
The units offer high temperature stability and are fitted with overtemperature and low level protection suitable for Safety Class III/FL (DIN 12876) for use with flammable fluids. A powerful pressure/suction pump ensures optimal circulation and temperature homogenity in the bath and also allows temperature control of externally attached applications using a pump adapter (accessory). The products are complimented by a range of accessories including inserts, variable adjustable platforms, bath covers, probes as well as hoses, temperature control fluids and various adapters.
The new multi-touch Pilot ONE controller offers a brilliant 5,7" touchscreen with graphic functions and a comfortable navigation menu. All important operating parameters and temperature values are neatly displayed on the touchscreen. Thanks to the new favourites menu, the one-click operation and the integrated technical glossary operation is very easy, just like on a smartphone. Integrated USB and Ethernet ports allow connection to a PC or network, e.g. for remote control or data transmission.
Maximum pump capacities: 27 L/min, 700 mbar (pressure), 22 L/min, 400 mbar (suction).
Maximum pump capacities:
Pilot ONE® (CC-E): 27 l/min, 700 mbar, 400 mbar suction
MPC: 20 l/min, 200 mbar, 180 mbar suction
Maximum pump capacities:
Pilot ONE® (CC-E): 27 l/min, 700 mbar, 400 mbar suction
MPC: 20 l/min, 0,2 bar, 0,18 bar suction
 : Safety Class III (FL) (DIN 12876) for use with flammable fluids.
 : Accessories included. There are also some optional accessories.
 : Note: The working temperature range of all models can be lowered with an optional cooling coil, connected to tap water or a chiller.
Product Family Options
Product Information
  • Type
    Temperature Range
    Temp. stability
    Heating capacity
    Capacity
    W×D×H
    Weight
    Availability
    Price
  • CC-104A
    (15)25...100 °C
    ±0,02 °C
    1,5 - 2,1 kW
    4 L
    147×235×330 mm
    6 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 360,00 €
    2 360,00 €
    1 * 1 ST
       2 360,00 €
  • CC-106A
    (15)25...100 °C
    ±0,02 °C
    1,5 - 2,1 kW
    6 L
    147×307×330 mm
    5 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 230,00 €
    2 230,00 €
    1 * 1 ST
       2 230,00 €
  • CC-108A
    (15)25...100 °C
    ±0,02 °C
    1,5 - 2,1 kW
    8 L
    147×407×330 mm
    6 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 240,00 €
    2 240,00 €
    1 * 1 ST
       2 240,00 €
  • CC-110A
    (15)25...100 °C
    ±0,02 °C
    1,5 - 2,1
    10 L
    147×507×330 mm
    6 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 310,00 €
    2 310,00 €
    1 * 1 ST
       2 310,00 €
  • CC-112A
    (15)25...100 °C
    ±0,02 °C
    1,5 - 2,1 kW
    12 L
    330×360×335
    8 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 490,00 €
    2 490,00 €
    1 * 1 ST
       2 490,00 €
  • CC-118A
    (15)25...100 °C
    ±0,02 °C
    1,5 - 2,1 kW
    18 L
    330×520×335 mm
    8 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 520,00 €
    2 520,00 €
    1 * 1 ST
       2 520,00 €