Fluorescent ruler, SP Bel-Art
BELAF133550001
- Description:Fluorescent ruler, Scienceware®
- Pk:1
Fluorescent ruler can be used in observing and photographing ethidium bromide stained nucleic acid agarose and acrylamide gels.
- Ruler fluorescences visibly upon illumination with ultraviolet light
- Measure of nucleic acid mobility or quantify the size of nucleic acid fragments
- Measurement allows for comparison of gels run at different times under the same conditions