ContactUsLinkComponent
Labelling tapes, autoclavable
Labelling tapes, autoclavable
Catalog # UTAP7160OR050-P3D
Supplier:  ULTRATAPE
Labelling tapes, autoclavable
Catalog # UTAP7160OR050-P3D
Supplier:  ULTRATAPE
Specifications
  • Width:
    6,3 mm
  • Description:
    Cleanroom labeling tape, with rubber adhesive
  • Pk:
    12 Roll
Specifications
About this item
Autoclavable vinyl tape with rubber adhesive.
  • Can be written on with a ball point pen
  • Suitable for medical and pharmaceutical applications