Order Entry
Austria
ContactUsLinkComponent
Urine sample containers, with screw cap
Urine sample containers, with screw cap
Catalog # SARS75.562.011
Supplier:  SARSTEDT
undefined
Urine sample containers, with screw cap
Catalog # SARS75.562.011
Supplier:  SARSTEDT

Specifications

  • Ext.Ø:
    62 mm
  • Packed:
    50/bag, 250/case
  • Capacity:
    100 ml
  • Description:
    Urine container with enclosed lid, brown
  • Height:
    72 mm
  • Pk:
    250

Specifications

About this item

PP, with brown screw cap

  • For safe light protection for urine samples, e.g. light sensitive cross links with deoxy pyridinoline assays.