ContactUsLinkComponent
Long handle ESD stick, Stat-Rite®
Long handle ESD stick, Stat-Rite®
Catalog # 115-1901
Supplier:  Texwipe
undefined
Long handle ESD stick, Stat-Rite®
Catalog # 115-1901
Supplier:  Texwipe
Specifications
  • Description:
    ESD sticks with micro tips
  • Length:
    56 mm
  • Pk:
    500
Specifications
About this item
This ESD-safe stick with micro tips is a double-ended cleaning and prepping tool.
  • Controls ESD without the need of carbons and ionic antistats
  • Made from Stat-Rite®, an inherently dissipative polymer
Designed for use at ESD-safe workstations.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.