ContactUsLinkComponent
PCR tubes, 0,5 ml, without cap, Axygen®
PCR tubes, 0,5 ml, without cap, Axygen®
Catalog # 732-1416
Supplier:  Corning
   Low Manufacturing Impact
undefined
PCR tubes, 0,5 ml, without cap, Axygen®
Catalog # 732-1416
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • Packed:
    500 tubes/pack
  • Capacity:
    0,5 ml
  • Colour:
    Clear
  • Description:
    Tubes, conical, without cap
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Pk:
    500
Specifications
About this item
PCR tube, made of PP, designed to fit standart 96 well blocks. Ultra-thin and consistent wall thickness allows precise thermal transfer and even insertion cap ensures a tight fit to prevent evaporation.
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Compatible with ABI-310 Instrument